Learn More
Bioworld Rappaport-Vassiliadis R10 Broth, 500 g
Catalog No. 50254459
Encompass_Preferred
Rappaport-Vassiliadis R10 BrothUsed for the selective enrichment of Salmonella sp. from meat products, feces, and sewage polluted waterpH @ 25°C : 5.1 ±2°CWorking Concentration: 26.6 g/LDehydrated Appearance: Pale green to green, homogeneous powderPrepared Appearance: blue, clear brothSoluble in warm waterContents (g/L):Tryptone- 4.54g Sodium Chloride-7.2gPotassium Dihydrogen Phosphate -1.45gMagnesium Chloride, Anhydrous-13.4gMalachite Green Oxalate -0.036g
Catalog No. 50-254-459 Supplier Bioworld Supplier No. 306230021
Rappaport-Vassiliadis R10 BrothUsed for the selective enrichment of Salmonella sp. from meat products, feces, and sewage polluted waterpH @ 25°C : 5.1 ±2°CWorking Concentration: 26.6 g/LDehydrated Appearance: Pale green to green, homogeneous powderPrepared Appearance: blue, clear brothSoluble in warm waterContents (g/L):Tryptone- 4.54g Sodium Chloride-7.2gPotassium Dihydrogen Phosphate -1.45gMagnesium Chloride, Anhydrous-13.4gMalachite Green Oxalate -0.036g