Learn More
AdipoGen Raptinal
SDP
Catalog No. 502562348
Encompass_Preferred
Chemical. CAS 1176-09-6. Formula C28H18O2. MW 386.4. Rapid apoptosis inducer that initiates intrinsic pathway caspase-dependent apoptosis within minutes in multiple cell lines and in vivo systems. Exerts anticancer activity in vivo, likely through the induction of apoptosis. Induces cell death against various cancer and non-cancerous cell lines with 24 hour IC50 values between 0.7-3.4µM indicating activity across a wide range of cell lines. Activates the intrinsic pathway and requires functional mitochondria for apoptosis induction. Treated cells did not undergo mitochondrial-permeability-transition-pore MPTP formation, suggesting a mitochondrial outer membrane permeabilization MOMP model of cytochrome c release.
Catalog No. 50-256-2348 Supplier AdipoGen Supplier No. AGCR12902M001
Chemical. CAS 1176-09-6. Formula C28H18O2. MW 386.4. Rapid apoptosis inducer that initiates intrinsic pathway caspase-dependent apoptosis within minutes in multiple cell lines and in vivo systems. Exerts anticancer activity in vivo, likely through the induction of apoptosis. Induces cell death against various cancer and non-cancerous cell lines with 24 hour IC50 values between 0.7-3.4µM indicating activity across a wide range of cell lines. Activates the intrinsic pathway and requires functional mitochondria for apoptosis induction. Treated cells did not undergo mitochondrial-permeability-transition-pore MPTP formation, suggesting a mitochondrial outer membrane permeabilization MOMP model of cytochrome c release.