Learn More
Enzo Life Sciences Raptor monoclonal antibody (10E10) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905765100
Encompass_Preferred
Clone: 10E10. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of Raptor. Species reactivity: Human, Mouse, Rat, Dog. Applications: WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-7042