Learn More
QuidelOrtho Rare Antisera
SDP
Catalog No. OK715130 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
3 mL
5 mL
Type:
Antisera Anti-Fya (Duffy)
Antisera Anti-Jka BioClone
Antisera Anti-Jkb BioClone
Antisera Anti-Kell
Antisera Anti-M (Rabbit)
Antisera Anti-S
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
OK715130 3 mL Antisera Anti-Fya (Duffy)
23-287-461 3 mL Antisera Anti-Jka BioClone
23-287-462 3 mL Antisera Anti-S
23-287-319 5 mL Antisera Anti-M (Rabbit)
23-287-322 5 mL Antisera Anti-Kell
23-287-323 3 mL Antisera Anti-Jkb BioClone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. OK715130 Supplier QuidelOrtho Supplier No. 715130
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Antisera are used as qualitative tests for recognition of antigens on human red blood cells
  • Each antisera is used to indicate the presence of specific antigens, which when present will cause clumping of normal human red blood cells
Specifications
Type Antisera Anti-Fya (Duffy)
For Use With (Application) For recognition of Antigens on Human red Blood cells
Quantity 3 mL