Learn More
Inorganic Ventures ICP MS RARE EARTH STD 125mL
SDP
Catalog No. 50723524
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Analyte: Ce, Er, Gd, La, Nd, Sm, Tb, Tm, Yb, Dy, Eu, Ho, Lu, Pr, Sc, Th, U, and Y 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-524 Supplier Inorganic Ventures Supplier No. CMS1125ML
Matrix: Nitric Acid (HNO3), Analyte: Ce, Er, Gd, La, Nd, Sm, Tb, Tm, Yb, Dy, Eu, Ho, Lu, Pr, Sc, Th, U, and Y 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search