Learn More
Sigma Aldrich Fine Chemicals Biosciences Rare earth extra power stir bars, 3/pk
Catalog No. NC1880828
Encompass
Elliptical, diam. 6 mm, L 10 mm. Stirring bars using rare earth magnets have significantly increased magnetic strength compared to those using Alnico magnets, but more importantly they are almost totally resistant to demagnetisation.
Catalog No. NC1880828 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6716573EA
Elliptical, diam. 6 mm, L 10 mm. Stirring bars using rare earth magnets have significantly increased magnetic strength compared to those using Alnico magnets, but more importantly they are almost totally resistant to demagnetisation.