Learn More
Research Products International Corp Rare Earth Cobalt Spinbar, 3 x 1/2
SDP
Catalog No. 502229627
Encompass
Reduce frequency of spinout in viscous solutions or when stirring at high speeds. The high magnetic field strength of Samarium Cobalt (Rare Earth) magnets can provide up to three times stronger coupling vs. Alnico magnetic stirring bars. Stronger coupling with the drive magnet keeps the stirring bar centered and rotating effectively. Octagon shape with pivot ring creates strong turbulence with minimal chattering and friction. Encapsulated in bright green, FDA grade, USP Class VI Teflon PTFE; easy to differentiate from other stirring bars. Autoclave safe.
Catalog No. 50-222-9627 Supplier Research Products International Corp Supplier No. 155423
Reduce frequency of spinout in viscous solutions or when stirring at high speeds. The high magnetic field strength of Samarium Cobalt (Rare Earth) magnets can provide up to three times stronger coupling vs. Alnico magnetic stirring bars. Stronger coupling with the drive magnet keeps the stirring bar centered and rotating effectively. Octagon shape with pivot ring creates strong turbulence with minimal chattering and friction. Encapsulated in bright green, FDA grade, USP Class VI Teflon PTFE; easy to differentiate from other stirring bars. Autoclave safe.