Learn More
Rarecyte RareCyte 25 um CytePicker II Picking Kit, 6-Pack
Supplier:  Rarecyte 241261025
Encompass
A pack of six of 25 um CytePicker II Needles with fifty CytePicker PCR Imaging Tubes. Compatible with CytePicker II which is compatible with the CyteFinder II instrument.
Catalog No. NC2069209