Learn More
Rarecyte RareCyte 40 um CytePicker II Picking Kit, 6-Pack
Catalog No. NC2069210
Encompass
A pack of six of 40 um CytePicker II Needles with fifty CytePicker PCR Imaging Tubes. Compatible with CytePicker II which is compatible with the CyteFinder II instrument.
Catalog No. NC2069210 Supplier Rarecyte Supplier No. 241261040
A pack of six of 40 um CytePicker II Needles with fifty CytePicker PCR Imaging Tubes. Compatible with CytePicker II which is compatible with the CyteFinder II instrument.