Learn More
Rarecyte RareCyte AccuCyte Blood Separation Kit, 10-Pack
Catalog No. NC2069162
Encompass
Individually packed components for processing ten blood samples. Components include: AccuCyte Blood Separation Tube with Cap, AccuCyte Float 1.055 - 1.058, AccuCyte Collector with Waste Tube, Isolation Tube with Septum Bottom, AccuCyte CyteSeal Brass Sealing Ring, AccuCyte Locking Clip and a grey CyteSpreader Blade.
Catalog No. NC2069162 Supplier Rarecyte Supplier No. 241288000
Individually packed components for processing ten blood samples. Components include: AccuCyte Blood Separation Tube with Cap, AccuCyte Float 1.055 - 1.058, AccuCyte Collector with Waste Tube, Isolation Tube with Septum Bottom, AccuCyte CyteSeal Brass Sealing Ring, AccuCyte Locking Clip and a grey CyteSpreader Blade.