Learn More
Rarecyte RareCyte AccuCyte Centrifuge Balances
Catalog No. NC2069213
Encompass
A set of two dissimilar AccuCyte centrifuge balances; to provide an equalizing counterweight on the centrifuge rotor when processing an odd number of tubes in a centrifuge. One type of centrifuge balance is used for the first spin and the other centrifuge balance is used for the second spin.
Catalog No. NC2069213 Supplier Rarecyte Supplier No. 241204000
A set of two dissimilar AccuCyte centrifuge balances; to provide an equalizing counterweight on the centrifuge rotor when processing an odd number of tubes in a centrifuge. One type of centrifuge balance is used for the first spin and the other centrifuge balance is used for the second spin.