Learn More
Rarecyte RareCyte AccuCyte Centrifuge Bucket Adapter, 2-Pack
Catalog No. NC2069216
Encompass
A set of two centrifuge bucket inserts. Each centrifuge bucket holds four AccuCyte tube adapters. These adapters are compatible with Beckman Allegra centrifuge with SX4750 rotor and recommend using it with this centrifuge.
Catalog No. NC2069216 Supplier Rarecyte Supplier No. 241203000
A set of two centrifuge bucket inserts. Each centrifuge bucket holds four AccuCyte tube adapters. These adapters are compatible with Beckman Allegra centrifuge with SX4750 rotor and recommend using it with this centrifuge.