Learn More
Rarecyte RareCyte AccuCyte CyteSeal Ring, 11-Pack
Catalog No. NC2069211
Encompass
A refill pack of eleven metal, deformable rings to be applied around the float and separation tube, sealing off the contents in a blood sample, below the ring.
Catalog No. NC2069211 Supplier Rarecyte Supplier No. 241118000
A refill pack of eleven metal, deformable rings to be applied around the float and separation tube, sealing off the contents in a blood sample, below the ring.