Learn More
Rarecyte RareCyte AccuCyte CyteSpreader Tray
Catalog No. NC2069212
Encompass
A reusable metal tray with a bubble level, level adjustment knobs and eight, grey, plastic bases to hold eight microscope slides. The plastic bases have a plastic guide to enable moving the CyteSpreader blade onto the microscope slide to help evenly distribute a monolayer of sample onto a microscope slide.
Catalog No. NC2069212 Supplier Rarecyte Supplier No. 221033001
A reusable metal tray with a bubble level, level adjustment knobs and eight, grey, plastic bases to hold eight microscope slides. The plastic bases have a plastic guide to enable moving the CyteSpreader blade onto the microscope slide to help evenly distribute a monolayer of sample onto a microscope slide.