Learn More
Rarecyte RareCyte CytePicker Chamber Slide Insert, 30-Pack
Catalog No. NC2069223
Encompass
A pack of thirty Inserts to be used with CytePicker Chamber Slides to prevent CytePicker Cell Adherence Solution from adhering to walls of chamber. Compatible with both, CyteFinder I and CyteFinder II instruments.
Catalog No. NC2069223 Supplier Rarecyte Supplier No. 241217000
A pack of thirty Inserts to be used with CytePicker Chamber Slides to prevent CytePicker Cell Adherence Solution from adhering to walls of chamber. Compatible with both, CyteFinder I and CyteFinder II instruments.