Learn More
Rarecyte RareCyte CytePicker Chamber Slide Centrifuge Adapter, 2-Pack
Supplier:  Rarecyte 241228000
Encompass
A set of two centrifuge adapters for holding CytePicker Chamber Slides. Compatible with Beckman Allegra centrifuge with SX4750 rotor and we recommend using it with this centrifuge. Compatible with both, CyteFinder I and Cytefinder II instruments.
Catalog No. NC2069230