Learn More
Rarecyte RareCyte CytePicker Chamber Slide Insert, 10-Pack
Supplier:  Rarecyte 241219000
Encompass
A pack of ten Inserts to be used with CytePicker Chamber Slides to prevent CytePicker Cell Adherence Solution from adhering to walls of chamber. Compatible with both, CyteFinder I and CyteFinder II instruments.
Catalog No. NC2069233