Learn More
Rarecyte RareCyte RarePlex CTC Panel Kit, Ventana (SYTOX)
Catalog No. NC2069173
Encompass
This is a 3-channel kit (Version: 0900-VB) that enables immunofluorescent detection of epithelial circulating tumor cells (CTCs). The kit contains a nuclear dye, a panel of antibodies and reagents for detection of CD45 (to exclude WBC), cytokeratin (CK), and epithelial cell adhesion molecule (EpCAM). Samples are stained by a Ventana DISCOVERY ULTRA automated staining system and imaged on a CyteFinderInstrument. The kit contains 250 slides, instructions and protocols for staining and scanning and guidelines for interpreting results.
Catalog No. NC2069173 Supplier Rarecyte Supplier No. 241245002
Add to Cart
Add to Cart
Request a Quote
This is a 3-channel kit (Version: 0900-VB) that enables immunofluorescent detection of epithelial circulating tumor cells (CTCs). The kit contains a nuclear dye, a panel of antibodies and reagents for detection of CD45 (to exclude WBC), cytokeratin (CK), and epithelial cell adhesion molecule (EpCAM). Samples are stained by a Ventana DISCOVERY ULTRA automated staining system and imaged on a CyteFinderInstrument. The kit contains 250 slides, instructions and protocols for staining and scanning and guidelines for interpreting results.