Learn More
Rarecyte RareCyte RarePlex CTC Panel Kit, Leica (SYTOX)
Catalog No. NC2069163
Encompass
This is a 3-channel kit (Version: 0900-LB) that enables immunofluorescent detection of epithelial circulating tumor cells (CTCs). The kit contains a nuclear dye, a panel of antibodies and reagents for the detection of CD45 (to exclude WBC), cytokeratin(CK) and epithelial cell adhesion molecule (EpCAM). Samples are stained with a Leica BOND RX automated staining system and are imaged on a CyteFinder instrument. The kit contains 200 slides, instructions and protocols for staining and scanning and guidelines for interpreting results.
Catalog No. NC2069163 Supplier Rarecyte Supplier No. 241245001
This is a 3-channel kit (Version: 0900-LB) that enables immunofluorescent detection of epithelial circulating tumor cells (CTCs). The kit contains a nuclear dye, a panel of antibodies and reagents for the detection of CD45 (to exclude WBC), cytokeratin(CK) and epithelial cell adhesion molecule (EpCAM). Samples are stained with a Leica BOND RX automated staining system and are imaged on a CyteFinder instrument. The kit contains 200 slides, instructions and protocols for staining and scanning and guidelines for interpreting results.