Learn More
Rarecyte RareCyte RarePlex Developer Kit: 405 nm, Leica
Catalog No. NC2069181
Encompass
The Developer Kit (Version: 405-A-L) enables detection of an additional custom biomarker in the 405 nm channel on a CyteFinder instrument. Samples are stained by a Leica BOND RX automated staining system and are imaged on a CyteFinder Instrument. This kit must be paired with a RarePlex base kit (0900-LB) to run on automated stainer. The kit contains reagents needed for detection of a single channel (405 nm) biomarker stained with a customer‑provided rabbit or mouse antibody, instructions and protocols for staining and scanning and guidelines for interpreting results.
Catalog No. NC2069181 Supplier Rarecyte Supplier No. 281001401
The Developer Kit (Version: 405-A-L) enables detection of an additional custom biomarker in the 405 nm channel on a CyteFinder instrument. Samples are stained by a Leica BOND RX automated staining system and are imaged on a CyteFinder Instrument. This kit must be paired with a RarePlex base kit (0900-LB) to run on automated stainer. The kit contains reagents needed for detection of a single channel (405 nm) biomarker stained with a customer‑provided rabbit or mouse antibody, instructions and protocols for staining and scanning and guidelines for interpreting results.