Learn More
Rarecyte RareCyte RarePlex Developer Kit: 405 nm, Ventana
Catalog No. NC2069175
Encompass
This Developer Kit (Version: 405-A-V) enables detection of an additional custom biomarker in the 405 nm channel on a CyteFinder instrument. Samples are stained by a Ventana DISCOVERY ULTRA automated staining system and are imaged on a CyteFinder Instrument. This kit must be paired with a RarePlex base kit (0900-VB) to run on automated stainer. The kit contains reagents needed for detection of a single channel (405 nm) biomarker stained with a customer‑provided rabbit or mouse antibody, instructions and protocols for staining and scanning and guidelines for interpreting results.
Catalog No. NC2069175 Supplier Rarecyte Supplier No. 281001402
This Developer Kit (Version: 405-A-V) enables detection of an additional custom biomarker in the 405 nm channel on a CyteFinder instrument. Samples are stained by a Ventana DISCOVERY ULTRA automated staining system and are imaged on a CyteFinder Instrument. This kit must be paired with a RarePlex base kit (0900-VB) to run on automated stainer. The kit contains reagents needed for detection of a single channel (405 nm) biomarker stained with a customer‑provided rabbit or mouse antibody, instructions and protocols for staining and scanning and guidelines for interpreting results.