Learn More
Aviva Systems Biology Corp RARG Antibody - middle region (ARP34364_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34364_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RARG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RARG. Alternate names: NR1B3, RARCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-55356