Learn More
Aviva Systems Biology Corp RARRES3 Antibody - middle region (ARP46474_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46474_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RARRES3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RARRES3. Alternate names: HRASLS4, MGC8906, RIG1, TIG3, PLA1/2-3ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59337