Learn More
Aviva Systems Biology Corp RARS Antibody - C-terminal region (ARP63097_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63097_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RARS. It was validated on Western Blot. . Alternate names: ArgRS, DALRD1, MGC8641ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-874