Learn More
Eurofins Discoverx raRsk1/MAPKAP Kinase 1a, active, 10 µg. (14-479)
Supplier:  Eurofins Discoverx 14479
Encompass
N-terminal 6xHis and HA (YPYDV PDYA) tagged recombinant, Rat full length RSK1 (residues 1-end). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8849