Learn More
Sigma Aldrich Fine Chemicals Biosciences Ras Assay Reagent Raf 1 RB
Catalog No. 502905932
Encompass
Product Source Human Ras Binding Domain (RBD residues 1-149) of Raf-1 expressed in E. coli.
Catalog No. 50-290-5932 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 14278
Product Source Human Ras Binding Domain (RBD residues 1-149) of Raf-1 expressed in E. coli.