Learn More
Cytoskeleton, Inc. Ras G-LISA Activation Assay Kit (Colorimetric Based) - 96 Assays
Supplier:  Cytoskeleton, Inc. BK131
Encompass
With the new Ras G-LISA kit you can now measure Ras activation from cell and tissue samples in less than 3 h.  G-LISA requires only 1-5% of the material needed for a conventional pull-down assay.  You will also be able to handle large sample numbers and generate quantitative results.
Catalog No. 50-646-244