Learn More
Enzo Life Sciences Ras inhibitory peptide (1mg)
SDP
Supplier:  Enzo Life Sciences ALX166001M001
Encompass_Preferred
The peptide sequence corresponds to amino acid residues 1149-1158 of the guanine nucleotide-releasing factor hSos1, which is essential for the control of Ras activity. Blocks the binding of hSos1 to human Grb2, a protein that binds to activated receptor tyrosine kinases. Alternative name: SOS-1 (human) (1149-1158). Formula: C53H91N19O11. MW: 1170.4. Purity: ≥97%. Appearance: White lyophilized powder. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7865