Learn More
Enzo Life Sciences Ras polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPGP001F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Ras. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Bovine. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7273