Learn More
Strem, An Ascensus Company CAS 2074610-05-0. 100mg. (R)-()-7-[4(S)-(Benzyl)oxazol-2-yl]-7-diphenylphosphino-2233-tetrahydro-11-spirobiindane min. 97% (RaS)-Ph-Bn-SIPHOX. MFCD17018764
SDP
Supplier:  Strem, An Ascensus Company 155186100MG
Encompass
CAS 2074610-05-0. 100mg. (R)-()-7-[4(S)-(Benzyl)oxazol-2-yl]-7-diphenylphosphino-2233-tetrahydro-11-spirobiindane min. 97% (RaS)-Ph-Bn-SIPHOX. MFCD17018764. Molecular Weight 563.67. Molecular Formula C39H34NOP. Color/form white solid Strem 15-5186. http//www.strem.com/catalog/v/15-5186/
Catalog No. 50-211-0767