Learn More
Medchemexpress LLC RAS/RAS-RAF-IN-1 | 2447039-81-6 | 99.8% | 807.98 | 1 MG
SDP
Catalog No. 5000365246
Encompass_Preferred
RAS/RAS-RAF-IN-1 is a potent RAS and RAS-RAF inhibitor that exhibits antitumor activity. It demonstrates a KD of 5.0 μM-15 μM for cyclophilin A (CYPA) binding affinity. This compound is intended for research use only.
  • Potent RAS and RAS-RAF inhibitor
  • Exhibits antitumor activity
  • Has a KD of 5.0 μM-15 μM for cyclophilin A (CYPA) binding affinity
  • Shows EC50 of 0.5 μM-5.0 μM for BRAF-GTP-KRAS-G12C disruption in H358 cells
  • Demonstrates IC50 of 1.0 μM-10 μM for pERK inhibition in H358 cells
  • For research use only
Catalog No. 50-003-65246 Supplier Medchemexpress LLC Supplier No. 1MGHY138294
RAS/RAS-RAF-IN-1 is a potent RAS and RAS-RAF inhibitor that exhibits antitumor activity. It demonstrates a KD of 5.0 μM-15 μM for cyclophilin A (CYPA) binding affinity. This compound is intended for research use only.
  • Potent RAS and RAS-RAF inhibitor
  • Exhibits antitumor activity
  • Has a KD of 5.0 μM-15 μM for cyclophilin A (CYPA) binding affinity
  • Shows EC50 of 0.5 μM-5.0 μM for BRAF-GTP-KRAS-G12C disruption in H358 cells
  • Demonstrates IC50 of 1.0 μM-10 μM for pERK inhibition in H358 cells
  • For research use only