Learn More
Medchemexpress LLC RAS/RAS-RAF-IN-1 | 2447039-81-6 | 99.8% | 807.98 | 1 ML
SDP
Catalog No. 5000348346
Encompass_Preferred
RAS/RAS-RAF-IN-1 is a potent inhibitor of both RAS and RAS-RAF pathways. It demonstrates a KD of 5.0 μM-15 μM for cyclophilin A (CYPA) binding affinity and exhibits antitumor activity.
  • Potent inhibitor of Ras and Raf.
  • Exhibits antitumor activity.
  • EC50 of 0.5 μM-5.0 μM for BRAF-GTP-KRAS-G12C disruption.
  • IC50 of 1.0 μM-10 μM for pERK inhibition in H358 cells.
Catalog No. 50-003-48346 Supplier Medchemexpress LLC Supplier No. HY13829410MM/1ML
RAS/RAS-RAF-IN-1 is a potent inhibitor of both RAS and RAS-RAF pathways. It demonstrates a KD of 5.0 μM-15 μM for cyclophilin A (CYPA) binding affinity and exhibits antitumor activity.
  • Potent inhibitor of Ras and Raf.
  • Exhibits antitumor activity.
  • EC50 of 0.5 μM-5.0 μM for BRAF-GTP-KRAS-G12C disruption.
  • IC50 of 1.0 μM-10 μM for pERK inhibition in H358 cells.