Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ras-related protein Rab-7a | 50 UG
SDP

Catalog No. 5000291165
Encompass_Preferred

Recombinant rat RAB7A protein with an N-terminal His tag, expressed in E. coli and supplied as a 0.22 μm filtered solution in PBS with 10% glycerol for research use in studies of endo-lysosomal trafficking and endosomal maturation. Product is low endotoxin and intended for biochemical and cell biology applications; store frozen and avoid repeated freeze-thaw.

  • Supplied as a solution in PBS with 10% glycerol.
  • N-terminal His tag for affinity applications.
  • Derived from rat sequence (accession P09527, M1-C207).
  • Expression system: E. coli.
  • Molecular weight approximately 25.7 kDa.
  • Endotoxin level <1 EU/μg (LAL method).
  • Store at -80°C for long-term stability; avoid repeated freeze-thaw.
  • Shipped on dry ice.

Catalog No. 50-002-91165 Supplier Medchemexpress LLC Supplier No. HYP75998A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat RAB7A protein with an N-terminal His tag, expressed in E. coli and supplied as a 0.22 μm filtered solution in PBS with 10% glycerol for research use in studies of endo-lysosomal trafficking and endosomal maturation. Product is low endotoxin and intended for biochemical and cell biology applications; store frozen and avoid repeated freeze-thaw.

  • Supplied as a solution in PBS with 10% glycerol.
  • N-terminal His tag for affinity applications.
  • Derived from rat sequence (accession P09527, M1-C207).
  • Expression system: E. coli.
  • Molecular weight approximately 25.7 kDa.
  • Endotoxin level <1 EU/μg (LAL method).
  • Store at -80°C for long-term stability; avoid repeated freeze-thaw.
  • Shipped on dry ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.