Learn More
Aviva Systems Biology Corp Rasa1 Antibody - C-terminal region (ARP42079_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42079_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rasa1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Gap, MGC7759, RasGAP, RasaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-974