Learn More
Apexbio Technology LLC Rasagiline Mesylate 161735-79-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000095072
Encompass_Preferred
Rasagiline mesylate (CAS 161735-79-1) is a selective irreversible inhibitor of monoamine oxidase type B (MAO-B) an enzyme responsible for the metabolic degradation of dopamine in the central nervous system By suppressing MAO-B activity rasagiline increases extracellular dopamine levels thereby modulating dopaminergic neurotransmission This mechanism underpins its use in research focused on neurodegenerative disorders particularly as a tool to investigate the pathophysiology and potential therapeutic interventions for idiopathic Parkinson s disease and related neurodegenerative conditions
Catalog No. 50-000-95072 Supplier Apexbio Technology LLC Supplier No. A43665.1
Rasagiline mesylate (CAS 161735-79-1) is a selective irreversible inhibitor of monoamine oxidase type B (MAO-B) an enzyme responsible for the metabolic degradation of dopamine in the central nervous system By suppressing MAO-B activity rasagiline increases extracellular dopamine levels thereby modulating dopaminergic neurotransmission This mechanism underpins its use in research focused on neurodegenerative disorders particularly as a tool to investigate the pathophysiology and potential therapeutic interventions for idiopathic Parkinson s disease and related neurodegenerative conditions