Learn More
Medchemexpress LLC Rasagiline (mesylate) | 161735-79-1 | 99.8% | 267.34 | 100 MG
SDP
Catalog No. 5000359767
Encompass_Preferred
Rasagiline mesylate is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor. It is also described as a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor
  • IC50s of 4.43 nM for rat brain MAO B activity
  • IC50s of 412 nM for rat brain MAO A activity
  • Click chemistry reagent
  • Capable of copper-catalyzed azide-alkyne cycloaddition
  • Increases proliferation rates of certain cell lines
  • Neuroprotective in a transgenic model of multiple system atrophy
Catalog No. 50-003-59767 Supplier Medchemexpress LLC Supplier No. HY14605100MG
Rasagiline mesylate is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor. It is also described as a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor
  • IC50s of 4.43 nM for rat brain MAO B activity
  • IC50s of 412 nM for rat brain MAO A activity
  • Click chemistry reagent
  • Capable of copper-catalyzed azide-alkyne cycloaddition
  • Increases proliferation rates of certain cell lines
  • Neuroprotective in a transgenic model of multiple system atrophy