Learn More
Medchemexpress LLC Rasagiline mesylate | 161735-79-1 | MFCD08460604 | 99.8% | 267.34 g/mol | C13H17NO3S | 5 MG
SDP
Catalog No. 5000222282
Encompass_Preferred
Rasagiline mesylate is a selective, irreversible monoamine oxidase (MAO) inhibitor provided for research applications. It is used in neuroscience and enzymatic assays to study MAO-B activity and is supplied in small solid and solution formats with analytical-grade purity.
  • Selective irreversible MAO inhibitor suitable for research assays.
  • High purity (99.82%) for analytical applications.
  • Available in small solid quantities and standard solutions.
  • Molecular weight 267.34 g/mol and formula C13H17NO3S.
  • Suitable for MAO-B inhibition and neuroscience studies.
Catalog No. 50-002-22282 Supplier Medchemexpress LLC Supplier No. HY146055MG
Rasagiline mesylate is a selective, irreversible monoamine oxidase (MAO) inhibitor provided for research applications. It is used in neuroscience and enzymatic assays to study MAO-B activity and is supplied in small solid and solution formats with analytical-grade purity.
  • Selective irreversible MAO inhibitor suitable for research assays.
  • High purity (99.82%) for analytical applications.
  • Available in small solid quantities and standard solutions.
  • Molecular weight 267.34 g/mol and formula C13H17NO3S.
  • Suitable for MAO-B inhibition and neuroscience studies.