Learn More
Medchemexpress LLC HY-14605 50mg Medchemexpress, Rasagiline (mesylate) CAS:161735-79-1 Purity:>98%
SDP
Catalog No. 502081622
Encompass_Preferred
Medchemexpress, HY-14605 50mg Rasagiline (mesylate) CAS:161735-79-1 Rasagiline (R-AGN1135) mesylate is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1622 Supplier Medchemexpress LLC Supplier No. HY1460550MG
Medchemexpress, HY-14605 50mg Rasagiline (mesylate) CAS:161735-79-1 Rasagiline (R-AGN1135) mesylate is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.