Learn More
Aviva Systems Biology Corp RASAL1 Antibody - C-terminal region (ARP62227_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62227_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RASAL1. It was validated on Western Blot. . Alternate names: RASALApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-287-621