Learn More
ABclonal Technology RASAL2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15120500UL
Encompass_Preferred
This gene encodes a protein that contains the GAP-related domain (GRD), a characteristic domain of GTPase-activating proteins (GAPs). GAPs function as activators of Ras superfamily of small GTPases. The protein encoded by this gene is able to complement the defective RasGAP function in a yeast system. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Catalog No. 50-001-07799