Learn More
Chemglass Life Sciences Rings, Raschig, 6 x 6mm, Distilling Column
SDP
Catalog No. CG128301
Encompass_Preferred
Rings are made of borosilicate glass and sold in one pound packages. 1lb of 6x6 rings is approx. 2304pcs which is roughly 391cm³1lb of 8x8 rings is approx. 1150pcs which is roughly 462cm³
Catalog No. CG128301 Supplier Chemglass Life Sciences Supplier No. CG128301
Rings are made of borosilicate glass and sold in one pound packages. 1lb of 6x6 rings is approx. 2304pcs which is roughly 391cm³1lb of 8x8 rings is approx. 1150pcs which is roughly 462cm³