Learn More
Research Products International Corp AeraSeal Replacement Rolls, Sterile, for use with SealMate Dispenser Kit, 2 Rolls per Package
SDP
Catalog No. 501977704
Encompass
Replacement rolls for use with the SealMate Dispenser Kit.
This adhesive film provides a leak tight seal on culture plates or 96 well assay plates. Unique micro-porous design allows uniform air and CO2 exchange between wells for optimum cell growth or bacterial cultivation. AeraSeal film minimizes cross-contamination between wells and reduces sample evaporation. Film can be easily pierced for sample recovery using a single or multi-channel pipettor.
Medical grade, non-cytotoxic adhesive has a thermal stability range of -20C to 80C. Removable tabs allow quick and easy positioning of film.
 AeraSeal and SealMate are Trademarks for Excel Scientific, Inc.
Catalog No. 50-197-7704 Supplier Research Products International Corp Supplier No. 202551
Replacement rolls for use with the SealMate Dispenser Kit.
This adhesive film provides a leak tight seal on culture plates or 96 well assay plates. Unique micro-porous design allows uniform air and CO2 exchange between wells for optimum cell growth or bacterial cultivation. AeraSeal film minimizes cross-contamination between wells and reduces sample evaporation. Film can be easily pierced for sample recovery using a single or multi-channel pipettor.
Medical grade, non-cytotoxic adhesive has a thermal stability range of -20C to 80C. Removable tabs allow quick and easy positioning of film.
 AeraSeal and SealMate are Trademarks for Excel Scientific, Inc.