Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Raseglurant hydrochloride | 757949-98-7 | 99.7% | 240.27 g·mol⁻1 | C15H13FN2 | 1 ML
SDP

Catalog No. 5000245165
Encompass_Preferred

Raseglurant hydrochloride is the hydrochloride salt of raseglurant (ADX-10059), a negative allosteric modulator of the metabotropic glutamate receptor mGluR5. It is provided as a 10 mM solution in DMSO for research use in pharmacology and in vitro studies; not for human use.

  • Hydrochloride salt of raseglurant, a selective mGluR5 negative allosteric modulator.
  • Supplied as a 10 mM solution in DMSO.
  • Quantity: 1 mL per vial.
  • High purity (~99.7%) suitable for research applications.
  • Molecular formula C15H13FN2; molecular weight ~240.27 g·mol⁻1.

Catalog No. 50-002-45165 Supplier Medchemexpress LLC Supplier No. HY10357410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Raseglurant hydrochloride is the hydrochloride salt of raseglurant (ADX-10059), a negative allosteric modulator of the metabotropic glutamate receptor mGluR5. It is provided as a 10 mM solution in DMSO for research use in pharmacology and in vitro studies; not for human use.

  • Hydrochloride salt of raseglurant, a selective mGluR5 negative allosteric modulator.
  • Supplied as a 10 mM solution in DMSO.
  • Quantity: 1 mL per vial.
  • High purity (~99.7%) suitable for research applications.
  • Molecular formula C15H13FN2; molecular weight ~240.27 g·mol⁻1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.