Learn More
AdipoGen Rasfonin
SDP
Supplier:  AdipoGen AGCN20173C500
Encompass_Preferred
Chemical. CAS 303156-68-5. Formula C25H38O6. MW 434.6. Isolated from Trichurus sp. Selective apoptosis inducer in ras-dependent cells. Autophagy Akt-dependent, apoptosis and necroptosis inducer in renal cancer cell lines. Induces death of ras-mutated pancreatic tumor Panc-1 cells in vitro and in vivo. Robust inhibitor of pancreatic cancers with the K-ras mutation. Reduced Sos1 expression and depressed Ras-MAPK activity.
Catalog No. 50-256-2644