Learn More
Cayman Chemical (–)-Rasfonin, Purity: ≥95%, CAS: 303156-68-5, 5 mg
SDP
Catalog No. 502937174
Encompass_Preferred
A fungal metabolite; inhibits concanavalin A- and LPS-induced proliferation of mouse splenocytes (IC50s = 0.7 and 0.5 μg/ml, respectively)
Catalog No. 50-293-7174 Supplier Cayman Chemical Supplier No. 275815 MG
A fungal metabolite; inhibits concanavalin A- and LPS-induced proliferation of mouse splenocytes (IC50s = 0.7 and 0.5 μg/ml, respectively)