Learn More
Aviva Systems Biology Corp RASGRF1 Antibody - middle region (ARP56515_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RASGRF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RASGRF1. Alternate names: CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-170