Learn More
Selleck Chemical LLC Raspberry ketone glucoside S3222-1mg
SDP
Catalog No. 503108281
Encompass_Preferred
Raspberry ketone glucoside (RKG) a natural product in raspberry fruit is an antioxidant that has the inhibitory effect on the melanin synthesis
Catalog No. 50-310-8281 Supplier Selleck Chemical LLC Supplier No. S32221MG
Raspberry ketone glucoside (RKG) a natural product in raspberry fruit is an antioxidant that has the inhibitory effect on the melanin synthesis