Learn More
Medchemexpress LLC Raspberry ketone (standard) | 5471-51-2 | MFCD00002394 | 164.20 | 50 MG
SDP
Catalog No. 5000416110
Encompass_Preferred
Raspberry ketone (Standard) is the analytical standard of Raspberry ketone. It is a major aromatic compound of red raspberry, widely used as a fragrance in cosmetics and as a flavoring agent in foodstuff. It also exhibits PPAR-α agonistic activity. This product is intended for research and analytical applications.
  • Analytical standard for Raspberry ketone
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Purity: 99.95%
Catalog No. 50-004-16110 Supplier Medchemexpress LLC Supplier No. HYN1426R50MG
Raspberry ketone (Standard) is the analytical standard of Raspberry ketone. It is a major aromatic compound of red raspberry, widely used as a fragrance in cosmetics and as a flavoring agent in foodstuff. It also exhibits PPAR-α agonistic activity. This product is intended for research and analytical applications.
  • Analytical standard for Raspberry ketone
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Purity: 99.95%