Learn More
Medchemexpress LLC Raspberry ketone (Standard) | 5471-51-2 | MFCD00004654 | 164.20 | 25 MG
SDP
Catalog No. 5000389486
Encompass_Preferred
Raspberry ketone (Standard) is the analytical standard of Raspberry ketone, intended for research and analytical applications. It is a major aromatic compound of red raspberry, widely used as a fragrance in cosmetics and as a flavoring agent in foodstuff; it also shows PPAR-α agonistic activity.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-89486 Supplier Medchemexpress LLC Supplier No. 25MGHYN1426R
Raspberry ketone (Standard) is the analytical standard of Raspberry ketone, intended for research and analytical applications. It is a major aromatic compound of red raspberry, widely used as a fragrance in cosmetics and as a flavoring agent in foodstuff; it also shows PPAR-α agonistic activity.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.