Learn More
Aviva Systems Biology Corp RASSF1 Antibody - C-terminal region (ARP59681_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RASSF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RASSF1. Alternate names: 123F2, NORE2A, RASSF1A, RDA32, REH3P21ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-776